Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Aprotinin, Analytical Grade, MP Biomedicals™

Catalog No. MP219038210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219038210 10 mg
MP219038250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219038210 Supplier MP Biomedicals™ Supplier No. 0219038210
Add to Cart
Add to Cart

Forms stable complexes with enzymes. Aprotinin, Analytical Grade, MP Biomedicals is a competitive serine proteinase inhibitor that works by blocking the active sites of enzymes. This binding is reversible with most aprotinin-protease complexes, dissociati

Aprotinin is found in bovine lymph nodes, lung, parotid gland, spleen, liver, pancreas, seminal vesicles, thyroid gland, kidney, mucous membranes of the trachea and esophagus, ovaries, heart, posterior pituitary and cartilage. It is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, kallkrein, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, and leukocyte and tissue proteinases. It does not inhibit thrombin. Uses include:

  • Small amounts can be added to tubes of drawn blood to enable laboratory measurement of rapidly degraded proteins, such as glucagon.
  • Can be labelled with fluorescein isothiocyanate for use as a fluorescent histochemical reagent for staining glycoconjugates (mucosubstances) that are rich in uronic or sialic acids. The conjugate retains its antiproteolytic and carbohydrate-binding properties.
  • Used in cell biology as an enzyme inhibitor to prevent protein degradation during lysis or homogenization of cells and tissues.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
CAS 9087-70-1
Molecular Formula C284H432N84O79S7
Synonym Pancreatic Basic Trypsin Inhibitor polypeptide; Antagosan; Antikrein; Trasylol; Zymofren; Antagosan; Anikrein; Antilysin(e); BPTI; Bayer A 128; Kallikrein-trypsin inactivator; Fosten; Iniprol; Kunitz protease inhibitor; Onquinin; Repulson; RP-9921; Ryker 52G; Triazinin; Zymofren
Content And Storage Store at 4°C
Donor Source Bovine Lung
Form Powder
For Use With (Application) Enzyme inhibitor
Quantity 10 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.