Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Aprotinin, from Bovine Lung

Catalog No. AAJ63039MC
Change view
Click to view available options
Quantity:
10mg
25mg
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63039MC 100mg
AAJ63039MA 10mg
AAJ63039MB 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63039MC Supplier Thermo Scientific Chemicals Supplier No. J63039MC
Add to Cart

missing translation for 'validMainframeMsg'

Aprotinin (iniprol, CAS # 9087-70-1) is a single polypeptide chain from bovine tissues with antifibrinolytic activities.

A potent protease inhibitor.Aprotinin acts as a reversible serine protease blocking and a binding agent. It is widely employed as an inhibitor of trypsin. It is also used as a proteolytic inhibitor in radioimmunoassays of polypeptide hormones. It is involved in the purification of urokinase, trypsin, and chymotrypsin on immobilized aprotinin. Further, it is used in the quantification of kallikrein activity in mixtures of esterases and proteases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Aprotinin is a 58-amino acid polypeptide chain found in lung tissue

• Aprotinin can suppress the systemic inflammatory response by reversibly inhibiting various esterases and proteases; it acts as a broad spectrum serine protease inhibitor

Application

• Aprotinin can be used as an enzyme inhibitor in laboratory studies on chymotrypsin, kallikrein, plasmin, and others

• Aprotinin is useful for the laboratory study of post-operative bleeding and wound healing

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protease Inhibition
Formulation Lyophilized powder
Quantity 100mg
Source Bovine lung
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Aprotinin, from Bovine Lung
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.