Learn More
Crescent Chemical Co Inc APROTININ FROM BOVINE LUNG
SDP
Catalog No. 50247305
Encompass_Preferred
Aprotinin from bovine lung lyophil. Purified inhibitor of trypsin, chymotrypsin, plasmin and especially kallikrein. For in vitro inhibition of fibrinolytic activity in blood samples (1). Size - 100MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 13718.
Catalog No. 50-247-305 Supplier Crescent Chemical Co Inc Supplier No. SE13718.03
Aprotinin from bovine lung lyophil. Purified inhibitor of trypsin, chymotrypsin, plasmin and especially kallikrein. For in vitro inhibition of fibrinolytic activity in blood samples (1). Size - 100MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 13718.