Learn More
Sigma Aldrich Fine Chemicals Biosciences Aprotinin from bovine lung
Catalog No. 502901128
Encompass
Trypsin inhibitor pancreas type from bovine lung. It is known as Pancreatic trypsin inhibitor (BPTI). Aprotinin also known as pancreatic trypsin inhibitor and trypsin-kallikrein inhibitor is found to be expressed in lungs spleen liver and pancreas. It is also found to be present in the free form in calf serum.
Catalog No. 50-290-1128 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10236624001
Trypsin inhibitor pancreas type from bovine lung. It is known as Pancreatic trypsin inhibitor (BPTI). Aprotinin also known as pancreatic trypsin inhibitor and trypsin-kallikrein inhibitor is found to be expressed in lungs spleen liver and pancreas. It is also found to be present in the free form in calf serum.