Learn More
Aprotinin From Bovine Lung
Description
A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
- Molecular Formula: C284H432N84O79S7
- MDLNumber: MFCD00130541
- CAS: 9087-70-1
Specifications
Specifications
| Concentration | 1 to 2 mg/mL (Soluble in 0.01 M HEPES) |
| For Use With (Application) | Protease Inhibition |
| Molecular Weight (g/mol) | 6511.44 |
| Name | Aprotinin |
| pH Range | 8 |
| Purification Method | Gel filtration |
| Quantity | 25 mg |
| Source | Bovine lung |
| Storage Requirements | 4°C |
| Packaging | Plastic bottle |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.