Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aprotinin From Bovine Lung

Catalog No. AAJ11388MC Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ11388MC 100 mg
AAJ11388MA 10 mg
AAJ11388MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ11388MC Supplier Thermo Scientific Supplier No. J11388MC
Add to Cart

missing translation for 'validMainframeMsg'

Useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Thermo Scientific™ Aprotinin From Bovine Lung is not effective against papain. | 6511.44 g/mol

A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
  • Molecular Formula: C284H432N84O79S7
  • MDLNumber: MFCD00130541
  • CAS: 9087-70-1
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 to 2 mg/mL (Soluble in 0.01 M HEPES)
For Use With (Application) Protease Inhibition
Molecular Weight (g/mol) 6511.44
Name Aprotinin
pH Range 8
Purification Method Gel filtration
Quantity 100 mg
Source Bovine lung
Storage Requirements 4°C
Packaging Plastic bottle
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.