Learn More
Aviva Systems Biology Corp APRT Antibody - N-terminal region (ARP58422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APRT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APRT. Alternate names: AMP, DKFZp686D13177, MGC125856, MGC125857, MGC129961ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Rat
Catalog No. 50-014-62302