Learn More
Research Products International Corp Ammonium Persulfate 100mg Tablets [APS], 100 Tablets
SDP
Catalog No. 50995775
Encompass_Preferred
Used as a catalyst for acrylamide gel polymerization.
Catalog No. 50-995-775 Supplier Research Products International Corp Supplier No. A22000100T
Used as a catalyst for acrylamide gel polymerization.