Learn More
Medchemexpress LLC HY-100627 10mg Medchemexpress, APS-2-79 CAS:2002381-25-9 Purity:>98%
SDP
Catalog No. 501931904
Encompass_Preferred
Medchemexpress, HY-100627 10mg APS-2-79 CAS:2002381-25-9 APS-2-79 behaves as a kinase suppressor of Ras ( KSR )-dependent antagonist of RAF-mediated MEK phosphorylation. APS-2-79 binds directly to KSR2 within the KSR2-MEK1 complex with an IC 50 of 120±23 nM for KSR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1904 Supplier Medchemexpress LLC Supplier No. HY10062710MG
Medchemexpress, HY-100627 10mg APS-2-79 CAS:2002381-25-9 APS-2-79 behaves as a kinase suppressor of Ras ( KSR )-dependent antagonist of RAF-mediated MEK phosphorylation. APS-2-79 binds directly to KSR2 within the KSR2-MEK1 complex with an IC 50 of 120±23 nM for KSR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.