Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apstatin trifluoroacetate | 99.4% | 573.56 g/mol | C25H34F3N5O7 | 5 MG
SDP

Catalog No. 5000264108
Encompass_Preferred

Apstatin TFA is the trifluoroacetate salt of apstatin, a potent aminopeptidase P (APP) inhibitor with reported Ki values of 2.6 μM (rat) and 0.64 μM (human). It has been used in preclinical studies showing cardioprotective effects and is supplied as a solid research compound for biochemical and pharmacological applications.

  • Potent aminopeptidase P inhibitor with reported Ki values of 2.6 μM (rat) and 0.64 μM (human).
  • Demonstrated cardioprotective effects in preclinical studies.
  • Supplied as a solid with reported purity of 99.43%.
  • Molecular formula C25H34F3N5O7 and molecular weight 573.56 g/mol.
  • Recommended storage: powder -80°C (2 years) or -20°C (1 year); protect from moisture.
  • Available in small research pack sizes suitable for preclinical experiments.

Catalog No. 50-002-64108 Supplier Medchemexpress LLC Supplier No. HY128090A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apstatin TFA is the trifluoroacetate salt of apstatin, a potent aminopeptidase P (APP) inhibitor with reported Ki values of 2.6 μM (rat) and 0.64 μM (human). It has been used in preclinical studies showing cardioprotective effects and is supplied as a solid research compound for biochemical and pharmacological applications.

  • Potent aminopeptidase P inhibitor with reported Ki values of 2.6 μM (rat) and 0.64 μM (human).
  • Demonstrated cardioprotective effects in preclinical studies.
  • Supplied as a solid with reported purity of 99.43%.
  • Molecular formula C25H34F3N5O7 and molecular weight 573.56 g/mol.
  • Recommended storage: powder -80°C (2 years) or -20°C (1 year); protect from moisture.
  • Available in small research pack sizes suitable for preclinical experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.