Learn More
Seracare Life Sciences Inc APSTOP SOLUTION (10X)
Catalog No. 5067457
Encompass_Preferred
10 X Stop solution for alkaline phosphatase-based ELISA specially formulated for use with BluePhos; Microwell Phosphatase Substrate (Cat. No. 50-88-00).
This item is not returnable. View return policy
Catalog No. 50-674-57 Supplier Seracare Life Sciences Inc Supplier No. 51500026
10 X Stop solution for alkaline phosphatase-based ELISA specially formulated for use with BluePhos; Microwell Phosphatase Substrate (Cat. No. 50-88-00).