Learn More
BINDER™ APT-COM™ 4 Series
SDP
Catalog No. 08111126 Shop All BINDER Inc. Products
Change view
Click to view available options
Includes:
4 BASIC-Edition Software
4 GLP-Edition Software
4 PROFESSIONAL-Edition Software
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
08-111-126 4 PROFESSIONAL-Edition Software
08-111-124 4 BASIC-Edition Software
08-111-125 4 GLP-Edition Software
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-111-126 Supplier BINDER™ Supplier No. 90530040
Add to Cart
Add to Cart
Multi Management Software provides management, logging, programming, and documentation options. With features ranging from GLP-compliant logging to convenient program creation and management of up to 100 units, the APT-COM™ 4 can be used in all industries and makes complex work processes much easier.
APT-COM™ 4 BASIC Edition:
  • Administration of up to five connected units
  • Log management (creating, deleting, archiving)
  • Documentation of measured values
  • Central overview of all units in both diagram and table form
  • Graphical presentation of measured values
  • Graphical/numerical program editor
  • Manual export of measured values (CSV/PDF file)
  • Multilingual user interface (German, English, French, Spanish, Italian)
  • Optional program execution via APT-COM™
  • Timer function
  • Import of data from APT-COM™ 3
APT-COM™ 4 PROFESSIONAL Edition:
All the same functions as the BASIC Edition and:
  • Administration of up to 100 connected units at the same time
  • Central overview of all units in a room plan
  • Email-based alarm system
  • Web server for displaying results
  • User administration for multiple users and authorizations
  • Automatic, time-managed export of measured values
  • Reports for further processing/archiving
  • Automatic/manual backup and recovery
APT-COM™ 4 GLP Edition
All the same functions as the PROFESSIONAL Edition and:
  • Can be validated according to 21 CFR Part 11 including validation documents
  • Documentation of measured values in a tamper-proof file in accordance with GLP/GMP guidelines on audit trails
  • Electronic signatures
For Use With: BINDER Chambers Series BD, BF, CB-S, CB, CBF, ED, FD, FED, FP, M, KB, KB ECO, KT, KBF, KBF-S, KBF-S ECO, KBF P, KBF LQC, KBW, KBWF, UFV, VD, VDL
Specifications
Product Type Data Collection Software
Includes 4 PROFESSIONAL-Edition Software
Description APT-COM™ 4 PROFESSIONAL-Edition - convenient unit and user management built on the BASIC edition. Suitable for networking up to 100 units.
For Use With (Application) Logging, Controlling, Monitoring
For Use With (Equipment) BINDER Chambers Series (BD, BF, CB-S, CB, CBF, ED, FD, FED, FP, M, KB, KB ECO, KT, KBF, KBF-S, KBF-S ECO, KBF P, KBF LQC, KBW, KBWF, UFV, VD, VDL)
Operating System Microsoft Windows 10 IoT 64-bit
Product Line APT-COM