Learn More
Meridian Life Science Aptamer Taq HS Poly Gly-Free, 20UL
SDP
Catalog No. 5000856326
Encompass_Preferred
Traditional hot start PCR methods that are antibody or chemically mediated require a long activation time at very high temperatures. In contrast, DNA aptamers disassociate from the enzyme (e.g. activate) at relatively lower temperatures (45 C), allowing for room temperature assay set-up, fast PCR reaction protocols and better assay specificity. Aptamer Taq HS (glycerol-free) is therefore suitable for difficult, multiplex PCR tests that require fast amplification whilst maintaining both high specificity and sensitivity.
Catalog No. 50-008-56326 Supplier Meridian Life Science Supplier No. MDX015002
Traditional hot start PCR methods that are antibody or chemically mediated require a long activation time at very high temperatures. In contrast, DNA aptamers disassociate from the enzyme (e.g. activate) at relatively lower temperatures (45 C), allowing for room temperature assay set-up, fast PCR reaction protocols and better assay specificity. Aptamer Taq HS (glycerol-free) is therefore suitable for difficult, multiplex PCR tests that require fast amplification whilst maintaining both high specificity and sensitivity.