Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aptiganel hydrochloride | 137160-11-3 | 99.8% | 339.86 g·mol⁻1 | C20H22ClN3 | 5 MG
SDP

Catalog No. 5000264904
Encompass_Preferred

Aptiganel hydrochloride is the hydrochloride salt of aptiganel, a non-competitive N-methyl-D-aspartate (NMDA) receptor antagonist with reported neuroprotective activity. It is provided as a purified research chemical for studies of excitotoxicity and NMDA receptor-mediated signaling.

  • Non-competitive NMDA receptor antagonist
  • Reported neuroprotective activity
  • Hydrochloride salt form
  • High purity (99.8%)
  • Suitable for in vitro and in vivo research
  • Molecular weight 339.86 g·mol⁻1

Catalog No. 50-002-64904 Supplier Medchemexpress LLC Supplier No. HY1100975MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aptiganel hydrochloride is the hydrochloride salt of aptiganel, a non-competitive N-methyl-D-aspartate (NMDA) receptor antagonist with reported neuroprotective activity. It is provided as a purified research chemical for studies of excitotoxicity and NMDA receptor-mediated signaling.

  • Non-competitive NMDA receptor antagonist
  • Reported neuroprotective activity
  • Hydrochloride salt form
  • High purity (99.8%)
  • Suitable for in vitro and in vivo research
  • Molecular weight 339.86 g·mol⁻1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.