Learn More
Medchemexpress LLC Aptiganel hydrochloride | 137160-11-3 | 99.9% | 339.86 | 100 MG
SDP
Catalog No. 5000344746
Encompass_Preferred
Aptiganel hydrochloride (Cerestat) is a non-competitive NMDA receptor antagonist with neuroprotective effect.
  • Non-competitive NMDA receptor antagonist
  • Neuroprotective effect
  • For research use only
Catalog No. 50-003-44746 Supplier Medchemexpress LLC Supplier No. HY11009710MG
Aptiganel hydrochloride (Cerestat) is a non-competitive NMDA receptor antagonist with neuroprotective effect.
  • Non-competitive NMDA receptor antagonist
  • Neuroprotective effect
  • For research use only