Learn More
Medchemexpress LLC Aptiganel hydrochloride | 137160-11-3 | 99.9% | 339.86 | 25 MG
SDP
Catalog No. 5000347677
Encompass_Preferred
Aptiganel hydrochloride (Cerestat) is a non-competitive NMDA receptor antagonist with a neuroprotective effect. This product is for research use only.
  • Acts as a non-competitive NMDA receptor antagonist.
  • Exhibits neuroprotective properties.
  • Intended for research use only.
Catalog No. 50-003-47677 Supplier Medchemexpress LLC Supplier No. 25MGHY110097
Aptiganel hydrochloride (Cerestat) is a non-competitive NMDA receptor antagonist with a neuroprotective effect. This product is for research use only.
  • Acts as a non-competitive NMDA receptor antagonist.
  • Exhibits neuroprotective properties.
  • Intended for research use only.