Learn More
Medchemexpress LLC Aptiganel hydrochloride (CNS 1102) | 137160-11-3 | 99.9% | 339.86 | 1 ML
SDP
Catalog No. 5000345013
Encompass_Preferred
Aptiganel hydrochloride is a non-competitive NMDA receptor antagonist that demonstrates a neuroprotective effect.
  • Acts as a non-competitive NMDA receptor antagonist.
  • Shows neuroprotective effect.
Catalog No. 50-003-45013 Supplier Medchemexpress LLC Supplier No. HY11009750MG
Aptiganel hydrochloride is a non-competitive NMDA receptor antagonist that demonstrates a neuroprotective effect.
  • Acts as a non-competitive NMDA receptor antagonist.
  • Shows neuroprotective effect.