Learn More
Selleck Chemical LLC APTO-253
SDP
Catalog No. 5000002996
Encompass_Preferred
APTO-253 (LOR-253 LT-253) inhibits c-Myc expression and selectively induces CDKN1A (p21) promotes G0-G1 cell-cycle arrest and triggers apoptosis in acute myeloid leukemia (AML) cells APTO-253 is also an inducer of KLF4 (Kr ppel-like factor 4)
Catalog No. 50-000-02996 Supplier Selleck Chemical LLC Supplier No. S69635MG
APTO-253 (LOR-253 LT-253) inhibits c-Myc expression and selectively induces CDKN1A (p21) promotes G0-G1 cell-cycle arrest and triggers apoptosis in acute myeloid leukemia (AML) cells APTO-253 is also an inducer of KLF4 (Kr ppel-like factor 4)