Learn More
Aviva Systems Biology Corp APTX Antibody - C-terminal region (ARP40014_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40014_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APTX. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human APTX. Alternate names: AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HITApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57263