Learn More
Aviva Systems Biology Corp APTX Antibody - N-terminal region (ARP32886_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32886_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APTX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APTX. Alternate names: AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HITApplicationWB, IHC. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54808