Learn More
Aviva Systems Biology Corp APTX Antibody - middle region (ARP32885_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32885_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APTX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human APTX. Alternate names: AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT, FLJ20157, MGC1072ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-014-54807