Learn More
Medchemexpress LLC APX2039 | 2342606-49-7 | 98.4% | 362.36 | 100 MG
SDP
Catalog No. 5000359491
Encompass_Preferred
APX2039 is an orally active and potent inhibitor of the fungal Gwt1 enzyme. It demonstrates extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*. It works by blocking the localization of GPI (glycosylphosphatidyl inositol)-anchored cell wall mannoproteins and can be used for Cryptococcal meningitis (CM) research.
  • Orally active and potent inhibitor of the fungal Gwt1 enzyme
  • Exhibits extremely potent anticryptococcal activity
  • Blocks localization of GPI-anchored cell wall mannoproteins
  • Useful for cryptococcal meningitis (CM) research
Catalog No. 50-003-59491 Supplier Medchemexpress LLC Supplier No. HY147110100MG
APX2039 is an orally active and potent inhibitor of the fungal Gwt1 enzyme. It demonstrates extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*. It works by blocking the localization of GPI (glycosylphosphatidyl inositol)-anchored cell wall mannoproteins and can be used for Cryptococcal meningitis (CM) research.
  • Orally active and potent inhibitor of the fungal Gwt1 enzyme
  • Exhibits extremely potent anticryptococcal activity
  • Blocks localization of GPI-anchored cell wall mannoproteins
  • Useful for cryptococcal meningitis (CM) research