Learn More
Medchemexpress LLC APX2039 | 2342606-49-7 | 98.4% | 362.36 | 5 MG
SDP
Catalog No. 5000361579
Encompass_Preferred
APX2039 is an orally active and potent inhibitor of the fungal Gwt1 enzyme, demonstrating extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*. It functions by blocking the localization of GPI (glycosylphosphatidyl inositol)-anchored cell wall mannoproteins and is suitable for Cryptococcal meningitis (CM) research.
  • Orally active and potent inhibitor of fungal Gwt1 enzyme.
  • Exhibits extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*.
  • Blocks the localization of GPI-anchored cell wall mannoproteins.
  • Applicable for Cryptococcal meningitis (CM) research.
Catalog No. 50-003-61579 Supplier Medchemexpress LLC Supplier No. HY1471105MG
APX2039 is an orally active and potent inhibitor of the fungal Gwt1 enzyme, demonstrating extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*. It functions by blocking the localization of GPI (glycosylphosphatidyl inositol)-anchored cell wall mannoproteins and is suitable for Cryptococcal meningitis (CM) research.
  • Orally active and potent inhibitor of fungal Gwt1 enzyme.
  • Exhibits extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*.
  • Blocks the localization of GPI-anchored cell wall mannoproteins.
  • Applicable for Cryptococcal meningitis (CM) research.