Learn More
Medchemexpress LLC APX2039 | 2342606-49-7 | 98.4% | 362.36 | 50 MG
SDP
Catalog No. 5000363587
Encompass_Preferred
APX2039 is an orally active and potent inhibitor of the fungal Gwt1 enzyme. It exhibits extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*, blocking the localization of glycosylphosphatidyl inositol (GPI)-anchored cell wall mannoproteins. This compound is suitable for Cryptococcal meningitis (CM) research.
  • Potent inhibitor of fungal Gwt1 enzyme
  • Exhibits anticryptococcal activity against *C. neoformans* and *C. gattii*
  • Blocks localization of GPI-anchored cell wall mannoproteins
  • Suitable for Cryptococcal meningitis (CM) research
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO
Catalog No. 50-003-63587 Supplier Medchemexpress LLC Supplier No. HY14711050MG
APX2039 is an orally active and potent inhibitor of the fungal Gwt1 enzyme. It exhibits extremely potent anticryptococcal activity against *C. neoformans* and *C. gattii*, blocking the localization of glycosylphosphatidyl inositol (GPI)-anchored cell wall mannoproteins. This compound is suitable for Cryptococcal meningitis (CM) research.
  • Potent inhibitor of fungal Gwt1 enzyme
  • Exhibits anticryptococcal activity against *C. neoformans* and *C. gattii*
  • Blocks localization of GPI-anchored cell wall mannoproteins
  • Suitable for Cryptococcal meningitis (CM) research
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO