Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC APX2039 | 2342606-49-7 | 98.4% | C20H15FN4O2 | 10MG
SDP

Catalog No. 5000258635
Encompass_Preferred

APX2039 is an orally active small-molecule inhibitor of the fungal Gwt1 enzyme with potent anticryptococcal activity in preclinical studies. Supplied as a research-grade solid, it is intended for in vitro and in vivo investigations and is not for human use.

  • Orally active Gwt1 enzyme inhibitor with potent anticryptococcal activity.
  • High purity solid (98.4%).
  • Molecular formula C20H15FN4O2; molecular weight 362.36 g·mol⁻¹.
  • Suitable for in vitro and in vivo preclinical research; not for human use.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-58635 Supplier Medchemexpress LLC Supplier No. HY14711010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APX2039 is an orally active small-molecule inhibitor of the fungal Gwt1 enzyme with potent anticryptococcal activity in preclinical studies. Supplied as a research-grade solid, it is intended for in vitro and in vivo investigations and is not for human use.

  • Orally active Gwt1 enzyme inhibitor with potent anticryptococcal activity.
  • High purity solid (98.4%).
  • Molecular formula C20H15FN4O2; molecular weight 362.36 g·mol⁻¹.
  • Suitable for in vitro and in vivo preclinical research; not for human use.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.