Learn More
New England Biolabs, Inc. Apyrase – 50 units
SDP
Catalog No. 501047494
Encompass
Apyrase (recombinant, E. coli) is a highly active ATP-diphosphohydrolase that catalyses the sequential hydrolysis of ATP to ADP and ADP to AMP releasing inorganic phosphate. It is a recombinant version of one of several isoforms of apyrase. It can also hydrolyse 5' tri- and diphosphate ribonucleosides and deoxyribonucleosides to their respective 5' monophosphates. Apyrase can catalyse the conversion of 5' triphosphorylated RNA to 5' monophosphorylated RNA.
Catalog No. 50-104-7494 Supplier New England Biolabs, Inc. Supplier No. M0398L
Apyrase (recombinant, E. coli) is a highly active ATP-diphosphohydrolase that catalyses the sequential hydrolysis of ATP to ADP and ADP to AMP releasing inorganic phosphate. It is a recombinant version of one of several isoforms of apyrase. It can also hydrolyse 5' tri- and diphosphate ribonucleosides and deoxyribonucleosides to their respective 5' monophosphates. Apyrase can catalyse the conversion of 5' triphosphorylated RNA to 5' monophosphorylated RNA.