Learn More
Sigma Aldrich Fine Chemicals Biosciences Apyrase from potato, 100 units
Catalog No. NC1596287
Encompass
Apyrase, also known as ATP-diphosphohydrolase, is expressed in wide variety of plant and animal tissues. The commercially accessible apyrase enzyme is isolated from potato tubers. recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder.
Catalog No. NC1596287 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A6237100UN
Apyrase, also known as ATP-diphosphohydrolase, is expressed in wide variety of plant and animal tissues. The commercially accessible apyrase enzyme is isolated from potato tubers. recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder.