Learn More
Sigma Aldrich Fine Chemicals Biosciences Apyrase from potatoes, 9000-95-7, MFCD00130542, 500UN
Catalog No. NC1568246
Encompass
Suitable for manufacturing of diagnostic kits and reagents, ATPase ≥200 units/mg protein. Synonym: Adenosine 5′-diphosphatase, Adenosine 5′-triphosphatase, Apyrase from potatoes. Apyrase is used to hydrolyze nucleoside triphosphates and diphosphates. For hydrolysis of organic di and triphosphates, the optimal pH is 6, and for inorganic substrates, the optimal pH is 5.1. Apyrase, from Sigma, has been used in inhibition studies of platelet-aggregation.
Catalog No. NC1568246 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SRE0019500UN
Suitable for manufacturing of diagnostic kits and reagents, ATPase ≥200 units/mg protein. Synonym: Adenosine 5′-diphosphatase, Adenosine 5′-triphosphatase, Apyrase from potatoes. Apyrase is used to hydrolyze nucleoside triphosphates and diphosphates. For hydrolysis of organic di and triphosphates, the optimal pH is 6, and for inorganic substrates, the optimal pH is 5.1. Apyrase, from Sigma, has been used in inhibition studies of platelet-aggregation.