Learn More
Cayman Chemical α-Pyrrolidinobutiothiophenone (hydrochloride), Purity: ≥98%, CAS: 2518265-88-6, 10 mg
SDP
Catalog No. 502971368
Encompass_Preferred
An analog of α-PBP that has a thiofuran group in place of the phenyl group
Catalog No. 50-297-1368 Supplier Cayman Chemical Supplier No. 1535810 MG
An analog of α-PBP that has a thiofuran group in place of the phenyl group