Learn More
Cayman Chemical aPyrrolDNoPNtIoTHOphenon 10mg
SDP
Catalog No. 502952931
Encompass_Preferred
An analog of the cathinone pyrovalerone in which the benzene ring has been replaced with a thiophene ring
Catalog No. 50-295-2931 Supplier Cayman Chemical Supplier No. 1418210 MG
An analog of the cathinone pyrovalerone in which the benzene ring has been replaced with a thiophene ring