Learn More
Medchemexpress LLC HY-100358 5mg Medchemexpress, AQ-13 dihydrochloride CAS:169815-40-1 Purity:>98%
SDP
Catalog No. 501931784
Encompass_Preferred
Medchemexpress, HY-100358 5mg AQ-13 dihydrochloride CAS:169815-40-1 AQ-13 dihydrochloride is an aminoquinoline antimalarial drug that is effective against chloroquine-resistant strains of Plasmodium falciparum. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1784 Supplier Medchemexpress LLC Supplier No. HY1003585MG
Medchemexpress, HY-100358 5mg AQ-13 dihydrochloride CAS:169815-40-1 AQ-13 dihydrochloride is an aminoquinoline antimalarial drug that is effective against chloroquine-resistant strains of Plasmodium falciparum. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.