Learn More
TARGETMOL CHEMICALS INC AQ-13 dihydrochloride 10MG
SDP
Catalog No. 502880583
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.AQ-13 dihydrochloride is an antimalarial drug. It is effective against drug-resistant strains of Plasmodium falciparum. Purity 99.88%
Catalog No. 50-288-0583 Supplier TARGETMOL CHEMICALS INC Supplier No. T1035610MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.AQ-13 dihydrochloride is an antimalarial drug. It is effective against drug-resistant strains of Plasmodium falciparum. Purity 99.88%