Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aq-13 dihydrochloride | 169815-40-1 | MFCD01938526 | 98.1% | 364.74 | C16H24Cl3N3 | 100 MG
SDP

Catalog No. 5000213279
Encompass_Preferred

AQ-13 dihydrochloride is an aminoquinoline antimalarial compound active against drug-resistant strains of Plasmodium falciparum, provided for research use. It is supplied as a light yellow solid and in solution formats, with molecular formula C16H24Cl3N3 and molecular weight 364.74.

  • Active against drug-resistant Plasmodium falciparum.
  • High purity (98.08%) appropriate for research applications.
  • Available as a light yellow solid and in solution formats.
  • Soluble in DMSO and water; may require ultrasonic agitation.
  • Stable when stored sealed away from moisture; in solvent, -80°C (6 months) or -20°C (1 month).
  • Molecular formula C16H24Cl3N3; molecular weight 364.74.

Catalog No. 50-002-13279 Supplier Medchemexpress LLC Supplier No. HY100358100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AQ-13 dihydrochloride is an aminoquinoline antimalarial compound active against drug-resistant strains of Plasmodium falciparum, provided for research use. It is supplied as a light yellow solid and in solution formats, with molecular formula C16H24Cl3N3 and molecular weight 364.74.

  • Active against drug-resistant Plasmodium falciparum.
  • High purity (98.08%) appropriate for research applications.
  • Available as a light yellow solid and in solution formats.
  • Soluble in DMSO and water; may require ultrasonic agitation.
  • Stable when stored sealed away from moisture; in solvent, -80°C (6 months) or -20°C (1 month).
  • Molecular formula C16H24Cl3N3; molecular weight 364.74.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.