Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AQ-13 dihydrochloride | 169815-40-1 | MFCD01938526 | 98.1% | 364.74 | C16H24Cl3N3 | 25 MG
SDP

Catalog No. 5000214335
Encompass_Preferred

AQ-13 dihydrochloride is an aminoquinoline antimalarial research compound used to study activity against Plasmodium falciparum, including drug-resistant strains. Supplied as the dihydrochloride salt for laboratory research use, it is provided as a solid with defined handling and storage recommendations.

  • Aminoquinoline-class compound effective against drug-resistant Plasmodium falciparum.
  • Dihydrochloride salt supplied as a light yellow to yellow solid.
  • High purity for research purposes (approximately 98.1%).
  • Molecular formula C16H24Cl3N3 and molecular weight 364.74.
  • Recommended storage: 4°C sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-14335 Supplier Medchemexpress LLC Supplier No. HY10035825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AQ-13 dihydrochloride is an aminoquinoline antimalarial research compound used to study activity against Plasmodium falciparum, including drug-resistant strains. Supplied as the dihydrochloride salt for laboratory research use, it is provided as a solid with defined handling and storage recommendations.

  • Aminoquinoline-class compound effective against drug-resistant Plasmodium falciparum.
  • Dihydrochloride salt supplied as a light yellow to yellow solid.
  • High purity for research purposes (approximately 98.1%).
  • Molecular formula C16H24Cl3N3 and molecular weight 364.74.
  • Recommended storage: 4°C sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.