Learn More
WELCH MATERIALS INC Ultisil AQ-C18, 10µm, 120Å, 4.6×250mm
Catalog No. NC2052211
Encompass_Preferred
Analytical Column, Ultisil AQ-C18, 10μm, 120Å, 4.6×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052211 Supplier WELCH MATERIALS INC Supplier No. H0020741043
Analytical Column, Ultisil AQ-C18, 10μm, 120Å, 4.6×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.