Learn More
WELCH MATERIALS INC Ultisil AQ-C18, 10µm, 50×250mm, Qty 1 each
Catalog No. NC2052603
Encompass_Preferred
Prep Column, Ultisil AQ-C18, 10µm, 50×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052603 Supplier WELCH MATERIALS INC Supplier No. H0261231210
Add to Cart
Add to Cart
Request a Quote
Prep Column, Ultisil AQ-C18, 10µm, 50×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.