Learn More
WELCH MATERIALS INC Ultisil AQ-C18, Cartridge: 3µm,120Å, 4.6×10mm, 10pcs/pk
Catalog No. NC2052531
Encompass_Preferred
Cartridge: Ultisil AQ-C18, 3µm,120Å, 4.6×10mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped. Matched HPLC Column ID: 3.0-8.0, Matched Guard Column Holder: 00808-01101 or 00808-01108, 10pcs/pk.
Catalog No. NC2052531 Supplier WELCH MATERIALS INC Supplier No. H008080300310
Cartridge: Ultisil AQ-C18, 3µm,120Å, 4.6×10mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped. Matched HPLC Column ID: 3.0-8.0, Matched Guard Column Holder: 00808-01101 or 00808-01108, 10pcs/pk.