Learn More
WELCH MATERIALS INC Ultisil AQ-C18, 5µm, 10×250mm
Catalog No. NC2052595
Encompass_Preferred
Prep Column, Ultisil AQ-C18, 5µm, 10×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.
Catalog No. NC2052595 Supplier WELCH MATERIALS INC Supplier No. H0261221102
Prep Column, Ultisil AQ-C18, 5µm, 10×250mm, pH range: 1.5-10.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 12%, Surface Area: 320m2/g, endcapped.