Learn More
Electron Microscopy Sciences Aqueous Glutaraldehyde 25% in Serum, EM Grade 10 ML
SDP
Catalog No. 5028594
Encompass_Preferred
Over the years, many of our valued customers have requested purchasing our aqueous Glutaraldehyde in serum vials. This packaging allows the user to puncture the special non-reactive seal with a hypodermic syringe and draw out the exact quantity of aqueous Glutaraldehyde needed without opening the container. This packaging assures freshness and purity of the aqueous Glutaraldehyde at all times.
Grade: EM Grade
Catalog No. 50-285-94 Supplier Electron Microscopy Sciences Supplier No. 16216
Over the years, many of our valued customers have requested purchasing our aqueous Glutaraldehyde in serum vials. This packaging allows the user to puncture the special non-reactive seal with a hypodermic syringe and draw out the exact quantity of aqueous Glutaraldehyde needed without opening the container. This packaging assures freshness and purity of the aqueous Glutaraldehyde at all times.
Grade: EM Grade