Learn More
Apexbio Technology LLC AQC 148757-94-2 25mg
SDP
Catalog No. 5000093439
Encompass_Preferred
AQC (148757-94-2) is a chemical reagent utilized for the derivatization of primary and secondary amines It reacts with amino groups to form stable fluorescent derivatives enabling sensitive detection and quantification of amino acids and related compounds Based on these properties AQC is widely applied in analytical biochemistry for high-performance liquid chromatography (HPLC)-based evaluation of amino acid composition protein sequencing and monitoring of biological processes such as enzymatic activities and metabolic pathway investigation
Catalog No. 50-000-93439 Supplier Apexbio Technology LLC Supplier No. A467725
AQC (148757-94-2) is a chemical reagent utilized for the derivatization of primary and secondary amines It reacts with amino groups to form stable fluorescent derivatives enabling sensitive detection and quantification of amino acids and related compounds Based on these properties AQC is widely applied in analytical biochemistry for high-performance liquid chromatography (HPLC)-based evaluation of amino acid composition protein sequencing and monitoring of biological processes such as enzymatic activities and metabolic pathway investigation