Learn More
Apexbio Technology LLC AQC 148757-94-2 50mg
SDP
Catalog No. 5000096332
Encompass_Preferred
AQC (148757-94-2) is a chemical reagent utilized for the derivatization of primary and secondary amines It reacts with amino groups to form stable fluorescent derivatives enabling sensitive detection and quantification of amino acids and related compounds Based on these properties AQC is widely applied in analytical biochemistry for high-performance liquid chromatography (HPLC)-based evaluation of amino acid composition protein sequencing and monitoring of biological processes such as enzymatic activities and metabolic pathway investigation
Catalog No. 50-000-96332 Supplier Apexbio Technology LLC Supplier No. A467750
AQC (148757-94-2) is a chemical reagent utilized for the derivatization of primary and secondary amines It reacts with amino groups to form stable fluorescent derivatives enabling sensitive detection and quantification of amino acids and related compounds Based on these properties AQC is widely applied in analytical biochemistry for high-performance liquid chromatography (HPLC)-based evaluation of amino acid composition protein sequencing and monitoring of biological processes such as enzymatic activities and metabolic pathway investigation