Learn More
Foxx Life Sciences CPC AseptiQuik C Connector, 3/8” Hose Barb Class VI Polycarbonate Coupling Body - HT, AQC17006HT, 5/PK
Catalog No. 5000572945
Encompass_Preferred
Manufactured by CPC, AseptiQuik C Connectors AQC17006HT provide quick and easy sterile connections, even in non-sterile environments. AseptiQuik C connectors’ “CLICK-PULL-TWIST” design enables users to transfer media easily with less risk of operator error. The connectors’ robust design provides reliable performance without the need for clamps, fixtures or tube welders. Biopharmaceutical manufacturers can make sterile connections with the quality and market availability they expect from the leader in single-use connection. Features CLICK-PULL-TWIST Design – Intuitive three-step connection process that reduces the risk of operator error. Enable simultaneous and secure removal of both membranes for a reliable sterile connection. Delivers repeatable, dependable performance without the need for additional hardware.Secures the final connection and prevents accidental disassembly. CPC Click Confirmation – Provides an audible “click” to confirm proper and complete assembly.
Catalog No. 50-005-72945 Supplier Foxx Life Sciences Supplier No. AQC17006HT5
Manufactured by CPC, AseptiQuik C Connectors AQC17006HT provide quick and easy sterile connections, even in non-sterile environments. AseptiQuik C connectors’ “CLICK-PULL-TWIST” design enables users to transfer media easily with less risk of operator error. The connectors’ robust design provides reliable performance without the need for clamps, fixtures or tube welders. Biopharmaceutical manufacturers can make sterile connections with the quality and market availability they expect from the leader in single-use connection. Features CLICK-PULL-TWIST Design – Intuitive three-step connection process that reduces the risk of operator error. Enable simultaneous and secure removal of both membranes for a reliable sterile connection. Delivers repeatable, dependable performance without the need for additional hardware.Secures the final connection and prevents accidental disassembly. CPC Click Confirmation – Provides an audible “click” to confirm proper and complete assembly.