Learn More
INFO AQL Inspector's Rule with Manual ANSI/ASQ Z1.4-2003
Catalog No. NC9955206
Encompass
Sampling Procedures And Tables For Inspection By Attributes. Softcover, 96 pages. Dimensions: 8 x 11. THIS IS THE CURRENT EDITION OF Z1.4. This standard is a very minor revision of ANSI/ASQ Z1.4-2003 (R2008), also referred to as ANSI/ASQ Z1.4-2008. It is an acceptance sampling system to be used with switching rules on a continuing stream of lots for AQL specified. It provides tightened, normal, and reduced plans to be applied for attributes inspection for percent nonconforming or nonconformities per 100 units.
Catalog No. NC9955206 Supplier INFO Supplier No. AQL ANSI ENG
Sampling Procedures And Tables For Inspection By Attributes. Softcover, 96 pages. Dimensions: 8 x 11. THIS IS THE CURRENT EDITION OF Z1.4. This standard is a very minor revision of ANSI/ASQ Z1.4-2003 (R2008), also referred to as ANSI/ASQ Z1.4-2008. It is an acceptance sampling system to be used with switching rules on a continuing stream of lots for AQL specified. It provides tightened, normal, and reduced plans to be applied for attributes inspection for percent nonconforming or nonconformities per 100 units.