Learn More
Molecular Devices Molecular Devices Objective, 40x S Fluor, 0.90 NA
Catalog No. 5000014663
Encompass_Preferred
High-performance 40x S Fluor microscope objective optimized for fluorescence imaging in high-content microscopy. It provides a high numerical aperture for improved resolution and light collection, and is designed for thin-bottom plate imaging.
  • 40x magnification for high-resolution imaging.
  • Numerical aperture 0.90 for enhanced light-gathering and resolution.
  • 0.3 mm working distance suitable for thin-bottom imaging.
  • Recommended thin bottom/cover thickness 0.17 mm.
  • Optimized for fluorescence applications.
Catalog No. 50-000-14663 Supplier Molecular Devices Supplier No. 163000197
Add to Cart
Add to Cart
Request a Quote
High-performance 40x S Fluor microscope objective optimized for fluorescence imaging in high-content microscopy. It provides a high numerical aperture for improved resolution and light collection, and is designed for thin-bottom plate imaging.
  • 40x magnification for high-resolution imaging.
  • Numerical aperture 0.90 for enhanced light-gathering and resolution.
  • 0.3 mm working distance suitable for thin-bottom imaging.
  • Recommended thin bottom/cover thickness 0.17 mm.
  • Optimized for fluorescence applications.