Learn More
Molecular Devices Molecular Devices Microplate Washer, 96- and 384-Well, Self-Contained, Interchangeable Heads
Catalog No. 5000014726
Encompass_Preferred
The Molecular Devices AquaMax 2000 Microplate Washer base unit is a self-contained instrument for precise, high-throughput washing of 96- and 384-well microplates. It supports interchangeable wash heads, multiple fluid inlets, and integration with automation systems for streamlined assays.
  • Self-contained operation with no external vacuum or computer required.
  • Configurable for 96- and 384-well plates, compatible with flat, U-, and V-bottom formats.
  • Interchangeable wash heads for rapid changeover and future scalability.
  • Multiple fluid inlet options to run several wash protocols without swapping bottles.
  • Fast, precise dispensing and aspiration with low residual volumes for reproducible assays.
  • Compact footprint and transportable weight of approximately 18.5 kg (41 lbs).
Catalog No. 50-000-14726 Supplier Molecular Devices Supplier No. 03105363
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices AquaMax 2000 Microplate Washer base unit is a self-contained instrument for precise, high-throughput washing of 96- and 384-well microplates. It supports interchangeable wash heads, multiple fluid inlets, and integration with automation systems for streamlined assays.
  • Self-contained operation with no external vacuum or computer required.
  • Configurable for 96- and 384-well plates, compatible with flat, U-, and V-bottom formats.
  • Interchangeable wash heads for rapid changeover and future scalability.
  • Multiple fluid inlet options to run several wash protocols without swapping bottles.
  • Fast, precise dispensing and aspiration with low residual volumes for reproducible assays.
  • Compact footprint and transportable weight of approximately 18.5 kg (41 lbs).