Learn More
Aviva Systems Biology Corp AQP10 Antibody - middle region (ARP50245_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50245_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AQP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human AQP10. Alternate names: AQPA_HUMANApplicationWB. Species reactivity: Human, Rat, Yeast
Catalog No. 50-014-60298