Learn More
Aviva Systems Biology Corp Aqp2 Antibody - middle region (ARP44295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Aqp2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AQP-CD, WCH-CD, cph, jpkApplicationWB, IF. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-285-134