Learn More
Aviva Systems Biology Corp AQP7 Antibody - C-terminal region (ARP32802_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32802_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AQP7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human AQP7. Alternate names: AQP9, AQP7L, AQPap, GLYCQTLApplicationWB. Species reactivity: Human
Catalog No. 50-014-54773