Learn More
Lauda Brinkmann Lp AQUA 90 BATH LIQUID 20L DRUM
Catalog No. 50931332
Encompass_Preferred
Bath Liquid Aqua 90 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 5 to 90oC, 20 L Drum This item is non returnable*
Catalog No. 50-931-332 Supplier Lauda Brinkmann Lp Supplier No. LZB 320
Bath Liquid Aqua 90 for LAUDA Circulators, Chillers and Thermostats, for use in applications between the temperature range of 5 to 90oC, 20 L Drum This item is non returnable*