Learn More
StatLab Aqua-Hold 2 Pap Pen
SDP
Catalog No. 22130516
Change view
Click to view available options
Aqua-Hold 2 Pap Pen
Catalog No.
22-130-516 Aqua-Hold 2 Pap Pen
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-516 Supplier StatLab Supplier No. 980402
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Draw red hydrophobic wells on slides to prevent waste of reagents and specimens
  • Once dry, the barrier is insoluble in alcohol and acetone
  • Barrier will wash off in 30 seconds with xylene or xylene substitutes
  • Each pen holds 13 mL of red liquid can make hundreds of easy-to-see red-tinted wells
  • 1 each
Specifications
Type Washable Marking Pen